Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Complement C4 (Human) ELISA Kit

Catalog No. 89100249 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-249 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-249 Supplier Abnova™ Supplier No. KA0467
Add to Cart
Add to Cart

Includes

  • Human Complement C4 Microplate: A 96-well polystyrene microplate coated with a polyclonal antibody against human complement C4
  • Sealing Tapes
  • Human Complement C4 Standard: Human complement C4 in a buffered protein base (lyophilized), 20μg
  • Biotinylated Human Complement C4: Lyophilized
  • MIX Diluent Concentrate (10x)
  • Wash Buffer Concentrate (20x)
  • Streptavidin-Peroxidase Conjugate (SP Conjugate): A 100-fold concentrate
  • Chromogen Substrate: A ready-to-use stabilized peroxidase chromogen substrate tetramethylbenzidine
  • Stop Solution: A 0.5N hydrochloric acid

Use to measure quantitatively human complement C4. A competitive enzyme immunoassay.

This assay employs a quantitative competitive enzyme immunoassay technique that measures human complement C4 in less than 3 hours

Specifications

Calibration Range 0.313 to 5μg/mL
Conjugate Biotin
Cross Reactivity Human
Description Complement C4 (Human) ELISA Kit is a competitive enzyme immunoassay for the quantitative measurement of human complement C4.
Detection Limit 0.3μg/mL
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status RUO
Sample Type Plasma, Serum
Sample Volume 25 μL
Packaging 96-well
Content And Storage Store components of the kit at 4°C or -20°C as described in the protocol.
For Use With (Application) Quantification
Product Type Complement C4 (Human) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.