Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Complement C4a des Arg (Human) ELISA Kit

Catalog No. 89100163 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-163 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-163 Supplier Abnova™ Supplier No. KA0308
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • Goat anti-Rabbit IgG Microtiter Plate, One Plate of 96 Wells
  • Human C4a des Arg EIA Conjugate, 6mL
  • Human C4a des Arg EIA Antibody, 6mL
  • Assay Buffer 10 Concentrate, 15mL
  • Wash Buffer Concentrate, 30mL
  • Human Complement C4a des Arg Standard, 0.5mL
  • Complement Reagent A, 15mL
  • Complement Reagent B, 30mL
  • pNpp Substrate, 20mL
  • Stop Solution, 6mL
  • Plate Sealers, 2 each

Use to measure quantitatively human C4a des Arg in plasma. A competitive immunoassay.

The kit uses a polyclonal antibody to human C4a des Arg to bind, in a competitive manner, the human C4a des Arg in the standard or sample or an alkaline phosphatase molecule which has human C4a des Arg attached to it

Specifications

Calibration Range 0.78 to 200ng/mL
Conjugate AP
Cross Reactivity Human
Description Complement C4a des Arg (Human) ELISA Kit is a competitive immunoassay for the quantitative determination of human C4a des Arg in plasma.
Detection Limit 0.76ng/mL
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Plasma (EDTA)
Sample Volume 100 μL
Sensitivity 0.76 ng/mL
Packaging 96-well
Content And Storage The standard and conjugate must be stored at -20°C; others store at 4°C.
For Use With (Application) Quantification
Product Type Complement C4a des Arg (Human) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.