Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Corticosterone ELISA Kit

Catalog No. 89100922 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-922 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-922 Supplier Abnova™ Supplier No. KA3385
Add to Cart
Add to Cart

Includes

  • Coated Clear 96 Well Plates: coated with donkey anti-sheep IgG
  • Corticosterone Standard: Corticosterone at 100,000 pg/mL in a special stabilizing solution
  • Corticosterone Antibody: sheep polyclonal antibody
  • Corticosterone Conjugate: A corticosterone-peroxidase conjugate
  • Assay Buffer Concentrate: A 5X concentrate
  • Dissociation Reagent: Dissociation Reagent is to be used only with Serum and Plasma samples
  • Wash Buffer Concentrate: A 20X concentrate
  • TMB Substrate
  • Stop Solution: A 1M solution of hydrochloric acid. CAUSTIC
  • Plate Sealer

Use to detect quantitatively of corticosterone in serum and plasma and in extracted fecal samples. Enzyme-linked immunosorbent assay.

This kit measures total corticosterone in serum and plasma and in extracted fecal samples

Specifications

Calibration Range 78.125 to 10,000pg/mL
Conjugate HRP
Description Corticosterone ELISA Kit is an enzyme-linked immunosorbent assay for the quantitative detection of corticosterone in serum and plasma and in extracted fecal samples.
Detection Limit 16.9pg/mL
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Dried Fecal Extract, Plasma (EDTA, Heparin), Serum,Tissue Culture Medium
Sample Volume 50 μL
Sensitivity 18.6 pg/mL
Packaging 96-well
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type Corticosterone ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.