Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Corticosterone (Pig) ELISA Kit

Catalog No. 89100598 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-598 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-598 Supplier Abnova™ Supplier No. KA2339
Add to Cart
Add to Cart

Includes

  • Antibody-coated microtiter wells, ready for use, 96 wells/plate
  • Reference Standard (0, 0.5, 1, 2.5, 5.0, 10, 25ng/mL), ready for use, 1 set
  • QC Controls: Low (1-2ng/mL) and high range (5-10ng/mL), ready for use, 1 set
  • HRP Conjugate Reagent, ready for use, 12mL
  • TMB Color Reagent, ready for use, 12mL
  • Stop solution (2N HCl), ready for use, 6mL
  • 20x Wash buffer, ready for use, 20mL
  • Sample diluent, ready for use, 25mL

Use to measure quantitatively corticosterone in Pig serum, plasma, urine. Immunoassay.

The Corticosterone Quantitative ELISA Test is based on a widely used immunoassay technique

Specifications

Calibration Range 0 to 20ng/mL
Conjugate HRP
Cross Reactivity Pig
Description Corticosterone (Pig) ELISA Kit is an immunoassay for the quantitative determination of corticosterone in Pig serum, plasma, urine.
Detection Limit 0.1ng/mL
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status RUO
Sample Type Plasma, Serum, Urine
Sample Volume 50 μL
Sensitivity 0.1 ng/mL
Packaging 96-well
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type Corticosterone (Pig) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.