Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Corticosterone (Rodent) ELISA Kit

Catalog No. 89100588 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-588 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-588 Supplier Abnova™ Supplier No. KA2329
Add to Cart
Add to Cart

Includes

  • Microtiter wells coated with Corticosterone specific antibody, 96 wells
  • Enzyme Conjugate, 12mL
  • QC Controls: QC-1 (1-2ng/mL) and QC-2 (5-10ng/mL), must be reconstitute using 1mL standard/sample diluent, 2 vials
  • TMB Color Reagent, 12mL
  • Stop Solution (2N HCl), 6mL
  • 20x Wash buffer, 20mL
  • Standard/Sample diluent, 20mL
  • Lyophilized standards: 0, 0.5, 1, 2, 5, 10, 20 ng per mL, must be reconstituted using 1mL standard/sample diluent, 1 set

Use to measure quantitatively corticosterone in rodent serum, plasma, urine. Immunoassay.

The Corticosterone Quantitative ELISA Test is based on a widely used immunoassay technique

Specifications

Calibration Range 0 to 20ng/mL
Conjugate HRP
Cross Reactivity Rodents
Description Corticosterone (Rodent) ELISA Kit is an immunoassay for the quantitative determination of corticosterone in rodent serum, plasma, urine.
Detection Limit 0.1ng/mL
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status RUO
Sample Type Plasma, Serum
Sample Volume 50 μL
Sensitivity 0.1 ng/mL
Packaging 96-well
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type Corticosterone (Rodent) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.