Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Cortisol (Canine) ELISA Kit

Catalog No. 89100547 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-547 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-547 Supplier Abnova™ Supplier No. KA2287
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • Microtiter wells coated with cortisol specific antibody, 96 wells
  • Enzyme Conjugate, 12mL
  • Lyophilized Standards, should be reconstituted in 1mL of standard/sample diluent (0, 1.0, 2.5, 5.0, 10, 50, 200ng/mL), 1 set
  • TMB Color Reagent (One-step ready for use), 12mL
  • Stop solution (2N HCl), 6mL
  • 20x Wash buffer, 20mL
  • Standard/Sample Diluent, 20mL

Use to measure quantitatively cortisol in canine serum, plasma. Immunoassay.

The cortisol quantitative assay is based on a widely used immunoassay technique

Specifications

Calibration Range 1 to 200ng/mL
Conjugate HRP
Cross Reactivity Dog
Description Cortisol (Canine) ELISA Kit is an immunoassay for the quantitative determination of cortisol in canine serum, plasma.
Detection Limit 1ng/mL
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status RUO
Sample Type Serum, Plasma
Sample Volume 25 μL
Sensitivity 1 ng/mL
Packaging 96-well
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type Cortisol (Canine) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.