Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Cortisol (Horse) Kit

Catalog No. 89100557 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-557 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-557 Supplier Abnova™ Supplier No. KA2298
Add to Cart
Add to Cart

Use to measure quantitatively cortisol in horse serum and plasma. Immunoassay.

Specifications

Calibration Range 0 to 200ng/mL
Conjugate HRP
Cross Reactivity Horse
Description Cortisol (Horse) ELISA Kit is an immunoassay for the quantitative determination of cortisol in horse serum and plasma.
Detection Method Colorimetric
Quantity 1 Kit
Sample Type Plasma, Serum, Urine
Sample Volume 25 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type Cortisol (Horse) ELISA Kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.