Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Cortisol (Sheep) ELISA Kit

Catalog No. 89100576 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
Plasma, Serum, Urine
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-100-576 Plasma, Serum, Urine
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-576 Supplier Abnova™ Supplier No. KA2317
Add to Cart
Add to Cart

Immunoassay for the quantitative determination of cortisol in sheep serum, plasma, urine

Specifications

Calibration Range 0 to 200ng/mL
Conjugate HRP
Cross Reactivity Sheep
Description Cortisol (Sheep) ELISA Kit is an immunoassay for the quantitative determination of cortisol in sheep serum, plasma, urine.
Detection Limit 1.0ng/mL
Detection Method Colorimetric
Includes 96 wells-Microtiter wells coated with cortisol specific antibody, HRPEnzyme Conjugate (12mL), Cortisol Standards 1 sets (0.5mL/vial), TMB Color reagent (12mL), Stop solution (6mL), Wash buffer 20X (25mL), Standard diluent (20mL)
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Plasma, Serum, Urine
Sample Volume 50 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type Cortisol (Sheep) ELISA kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.