Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Cotinine ELISA Kit

Catalog No. 89024471 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
96 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-024-471 96 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-024-471 Supplier Abnova™ Supplier No. KA0930

Includes

  • Microwell coated with polyclonal Ab to Cotinine, 12 x 8 x 1
  • Standard set (ready to use), 0.5mL
  • Cotinine HRP enzyme conjugate (ready to use), 12mL
  • TMB substrate (ready to use), 12mL
  • Stop solution (ready to use), 12mL

For the quantitative measurement of Cotinine in Serum and Urine

Specifications

Calibration Range 5 to 100ng/mL
Conjugate HRP
Description Cotinine ELISA Kit is used for the measurement of Cotinine.
Detection Limit 1ng/mL
Detection Method Colorimetric
Includes 12 x 8 x1 Microwell coated with Polyclonal Ab to Cotinine, 0.5mL standard set, 12mL Cotinine HRP Enzyme Conjugate, 12mL TMB Substrate, 12mL stop solution
Quantity 96 Assays
Sample Type Serum, Urine
Sample Volume 10 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Qualitative
Product Type Cotinine ELISA Kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.