Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Cotinine (Mouse/Rat) ELISA Kit

Catalog No. 89100526 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-526 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-526 Supplier Abnova™ Supplier No. KA2264

Includes

  • Microwell coated with polyclonal Ab to Cotinine, 12 x 8 x 1
  • Standard Set (ready to use), 0.5mL x 6
  • Cotinine HRP Enzyme Conjugate (ready to use), 12mL
  • TMB Substrate: (ready to use), 12mL
  • Stop Solution: (ready to use), 12mL

Measure Cotinine in mouse or rat serum or urine.

The Cotinine kit is a solid phase competitive ELISA

Specifications

Calibration Range 5 to 100ng/mL
Conjugate HRP
Cross Reactivity Mouse, Rat
Description Cotinine (Mouse/Rat) ELISA Kit is intended for the measurement of Cotinine in mouse or rat serum or urine.
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Serum, Urine
Sample Volume 10 μL
Packaging 96-well
Content And Storage Store the kit at 4°C
For Use With (Application) Qualitative
Product Type Cotinine (Mouse/Rat) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.