Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ CP (Human) ELISA Kit

Catalog No. 89100247 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
CCS, Milk, Saliva, Urine
Plasma, Serum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-100-247 Plasma, Serum
89-100-246 CCS, Milk, Saliva, Urine
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-100-247 Supplier Abnova™ Supplier No. KA0469

missing translation for 'validMainframeMsg'

Includes

  • Human Ceruloplasmin Microplate
  • Sealing Tapes: Precut, pressure-sensitive sealing tapes that can be cut to fit the format of the individual assay
  • Human Ceruloplasmin Standard: Human ceruloplasmin in a buffered protein base (lyophilized), 80μg
  • Biotinylated Human Ceruloplasmin: Lyophilized
  • MIX Diluent Concentrate (10x)
  • Wash Buffer Concentrate (20x)
  • Streptavidin-Peroxidase Conjugate (SP Conjugate): A 100-fold concentrate
  • Chromogen Substrate: A ready-to-use stabilized peroxidase chromogen substrate tetramethylbenzidine
  • Stop Solution: A 0.5N hydrochloric acid

This assay employs a quantitative competitive enzyme immunoassay technique that measures human ceruloplasmin in less than 3 hours

Specifications

Calibration Range 0.63 to 40ng/mL
Conjugate Biotin
Cross Reactivity Human
Detection Limit 0.6μg/mL
Detection Method Colorimetric
Includes Antibody coated microwells (96 wells), Human Ceruloplasmin Standard (20μg), Biotinylated Ceruloplasmin (1 vial), MIX Diluent Concentrate (10x): 30mL, Wash Buffer Concentrate (20x): 30mL, Streptavidin-Peroxidase Conjugate (SP Conjugate): 80, Chromogen Substrate (8mL), Stop solution (12mL)
Regulatory Status For research use only (RUO)
Sample Type Plasma, Serum
Sample Volume 25 μL
Packaging 96-well
Content And Storage -20°C (Aliquot to avoid repeated freezing and thawing)
For Use With (Application) Quantitative measurement of human CP in plasma and serum
Product Type ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.