Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ CRP (Human) ELISA Kit

Catalog No. 89022717 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-022-717 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-022-717 Supplier Abnova™ Supplier No. KA0238
Add to Cart
Add to Cart

Includes

  • Microtiter wells coated with mouse monoclonal anti-CRP 96 wells
  • Reference Standard Set Contains 0, 0.005, 0.010, 0.025, 0.050 and 0.100mg/l CRP in serum based buffer with preservatives 1mL each
  • hsCRP Sample Diluent Contains phosphate buffer-BSA solution with preservatives. 50mL/vial
  • CRP Enzyme Conjugate Reagent Contains goat anti-CRP conjugated to horseradish peroxidase with preservatives. 12mL
  • TMB Reagent (One-Step) 11mL
  • Stop Solution (1N HCl) 11mL

Sandwich Enzyme Immunoassay for the quantitative measurement of C- reactive Protein

Specifications

Calibration Range 0 to 0.1mg/L
Conjugate HRP
Cross Reactivity Human
Description CRP (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of C- reactive protein.
Detection Limit 0.1mg/L
Detection Method Colorimetric
Includes 96-well microtiter well coated with Mouse Monoclonal Anti-CRP, 1mL each reference standard set contains 0, 0.005, 0.010, 0.025, 0.050 and 0.100 mg/l, 50mL sample diluent, 12mL Enzyme Conjugate reagent, 11mL TMB reagent (One-Step), 11mL stop solution (1N HCl)
Quantity 1 Kit
Sample Type Serum
Sample Volume 10 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type CRP (Human) ELISA Kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.