Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ CRP (Monkey), HRP, ELISA Kit

Catalog No. 89100642 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-642 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-642 Supplier Abnova™ Supplier No. KA2447
Add to Cart
Add to Cart

Includes

  • anti-monkey CRP antibody coated microtiter plate 96 wells (provided as 12 detachable strips of 8)
  • Enzyme Conjugate Reagent 11mL
  • Reference standard containing CRP (concentration and dilution instructions are listed on the vial label)A 1 vial
  • 10X Diluent 25mL
  • 20x Wash Solution 50mL
  • TMB Reagent (One-Step) 11mL
  • Stop Solution (1N HCl) 11mL

For quantitative measurement of IgM in plasma and serum

Specifications

Calibration Range 1.17 to 75ng/mL
Conjugate HRP
Cross Reactivity Monkey
Description CRP (Monkey) ELISA Kit is for the quantitative measurement of CRP in serum.
Detection Limit 1.17 to 75 ng/mL
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Serum
Sample Volume 100 μL
Content And Storage Store lyophilized standard at -20°C. Store all other components at 4°C.
For Use With (Application) Quantification
Product Type CRP (Monkey) ELISA Kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.