Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Crp (Rat) ELISA Kit

Catalog No. 89028157 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-157 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-157 Supplier Abnova™ Supplier No. KA1035
Add to Cart
Add to Cart

Sandwich Enzyme Immunoassay for the quantitative measurement of Rat Crp in Urine and cell culture supernatants

Specifications

Calibration Range 0.78 to 50ng/mL
Conjugate HRP
Cross Reactivity Rat
Description Crp (Rat) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of rat Crp in urine and cell culture supernatants.
Detection Limit 0.7ng/mL
Detection Method Colorimetric
Includes 96-well Antibody coated microwell, 100ng Rat Crp standard, 1 vial (70X) Biotinylated Crp Antibody, 30mL 1 vial (10X) EIA diluent concentrate, 30mL Wash Buffer 20X, 90 Streptavidin-Peroxidase Conjugate (SP Conjugate), 8mL Chromogen Substrate, 12mL stop solution
Quantity 1 Kit
Sample Type Cell Culture Supernatant, Plasma, Serum, Urine
Sample Volume 50 μL
Content And Storage Store components of the kit at 4°C or -20°C as described in the protocol.
For Use With (Application) Quantification
Product Type CRP (Rat) ELISA kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.