Learn More
Specifications
Specifications
| Conjugate | HRP |
| Cross Reactivity | Human |
| Description | Cryptosporidium spp. ELISA Kit is an enzyme linked immunosorbent assay for the detection of Cryptosporidium spp. cells in human stool. |
| Detection Method | Colorimetric |
| Includes | 96-well break apart microtiter plate coated with Anti-Cryptosporidium species Polyclonal Antibodies, 1 bottle, 100mL (20X) concentrated Wash Buffer, 1 bottle, 60mL/1 bottle, 100mL Stool Diluent (Blue), 1 bottle, 12mL/1 bottle, 16mL HRP Conjugate (Green), |
| Quantity | 96 Assays |
| Sample Type | Stool |
| Sample Volume | 100 μL |
| Content And Storage | Store the kit at 4°C. Do not freeze. |
| For Use With (Application) | Quantification |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.