Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ CST3 (Human) ELISA Kit

Catalog No. 89100008 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
Body Fluid, CCS, Plasma, Serum, Tissue Lysates
CSF, Plasma (Citrate, EDTA, Heparin), Serum, Urine
Plasma (EDTA, Heparin), Serum, Tissue Culture Medium, Urine
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-100-008 CSF, Plasma (Citrate, EDTA, Heparin), Serum, Urine
89-100-379 Body Fluid, CCS, Plasma, Serum, Tissue Lysates
89-100-921 Plasma (EDTA, Heparin), Serum, Tissue Culture Medium, Urine
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 89-100-008 Supplier Abnova™ Supplier No. KA0022
Add to Cart
Add to Cart

Includes

  • Antigen Coated Microtiter Strips, 96 wells
  • Conjugate Solution Conc. (50x), 0.26mL
  • Conjugate Diluent (ready to use), 13mL
  • Set of Standards (concentrated), 0.1mL x 6
  • Quality Control HIGH (concentrated), 0.1mL
  • Quality Control LOW (concentrated), 0.1mL
  • Dilution Buffer Conc. (10 x), 10mL
  • Wash Solution Conc. (10 x), 100mL
  • Substrate Solution (ready to use), 13mL
  • Stop Solution (ready to use), 13mL

  • The kit measures total cystatin C in serum, plasma (EDTA, citrate, heparin), urine and cerebrospinal fluid
  • Quality Controls are human serum or human urine native protein based. No animal sera are used
  • Standard is purified native protein based
  • Components of the kit are provided ready to use or concentrated
  • Convenient for automatization

Specifications

Calibration Range 200 to 10000ng/mL
Conjugate HRP
Cross Reactivity Human, Monkey
Description CST3 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human cystatin C.
Detection Limit 0.25ng/mL
Detection Method Colorimetric
Includes Antibody coated microtiter strips (96 wells), Conjugate solution conc. 50X (0.26mL), Conjugate diluent (13mL), Set of standards (6 x 0.1mL), Quality control HIGH (0.1mL), Quality control Low (0.1mL), Dilution buffer Conc. (10x): 10mL, Wash solution conc. (10x): 100mL, Substrate solution (13mL), Stop solution 13mL
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type CSF, Plasma (Citrate, EDTA, Heparin), Serum, Urine
Sample Volume 100 μL
Sensitivity 0.25 ng/mL
Total Assay Time <2 hr.
Packaging 96-well
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type CST3 (Human) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.