Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ CST3 (Human) ELISA Kit

Catalog No. 89100921 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
Body Fluid, CCS, Plasma, Serum, Tissue Lysates
CSF, Plasma (Citrate, EDTA, Heparin), Serum, Urine
Plasma (EDTA, Heparin), Serum, Tissue Culture Medium, Urine
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-100-921 Plasma (EDTA, Heparin), Serum, Tissue Culture Medium, Urine
89-100-008 CSF, Plasma (Citrate, EDTA, Heparin), Serum, Urine
89-100-379 Body Fluid, CCS, Plasma, Serum, Tissue Lysates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 89-100-921 Supplier Abnova™ Supplier No. KA3384
Only null left
Add to Cart
Add to Cart

Includes

  • Clear Coated 96 Well Plate: Clear plastic microplate with break-apart strips coated with mouse anti-human Cystatin C, 1 plate
  • Cystatin C Standard: A stock solution of native human Cystatin C at 400ng/mL, 60μL
  • Cystatin C Conjugate: A monoclonal antibody to Cystatin C labeled with peroxidase, 5mL
  • Assay Buffer Concentrate: A 5X concentrate, 28mL
  • Wash Buffer Concentrate: A 20X concentrate, 30mL
  • TMB Substrate, 5mL
  • Stop Solution: A 1N hydrochloric acid solution. Caustic, 5mL
  • Plate Sealer, 1 each

The CST3 (Human) ELISA Kit is designed to quantitatively measure human Cystatin C present in biological samples and tissue culture media

Specifications

Calibration Range 0 to 10ng/mL
Conjugate HRP
Cross Reactivity Human
Description CST3 (Human) ELISA Kit is an enzyme-linked immunosorbent assay for the quantitative detection of human CST3 in biological samples and tissue culture media.
Detection Limit 0.171ng/mL
Detection Method Colorimetric
Includes Clear Coated 96 Well Plate: Clear plastic microplate with break-apart strips coated with mouse anti-human Cystatin C (1 plate), Cystatin C Standard: A stock solution of native human Cystatin C at 400ng/mL (60μL), Cystatin C Conjugate: A monoclonal antibody to Cystatin C labeled with peroxidase (5mL), Assay Buffer Concentrate: A 5X concentrate that should be diluted with deionized or distilled water (28mL), Wash Buffer Concentrate: A 20X concentrate that should be diluted with deionized or distilled water (30mL), TMB substrate (5mL), Stop Solution: A 1N hydrochloric acid solution. Caustic (5mL), Plate sealer (1 each)
Regulatory Status For research use only (RUO)
Sample Type Plasma (EDTA, Heparin), Serum, Tissue Culture Medium, Urine
Sample Volume 50 μL
Sensitivity 0.058 ng/mL
Packaging 96-well
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type CST3 (Human) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.