Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ CTF1 (Human) ELISA Kit

Catalog No. 89100730 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-730 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-730 Supplier Abnova™ Supplier No. KA3023
Add to Cart
Add to Cart

A sandwich enzyme immunoassay for the quantitative measurement of human CTF1 in sera, body fluids, tissue lysates or cell culture supernates

  • The protein encoded by this gene is a secreted cytokine that induces cardiac myocyte hypertrophy in vitro
  • It has been shown to bind and activate the ILST/gp130 receptor
  • Two transcript variants encoding different isoforms have been found for this gene

For research use only. Not for diagnostic and clinical use.

Specifications

Calibration Range 31.2 to 2000pg/mL
Description CTF1 (Human) ELISA Kit is intended for the quantitative measurement of human CTF1.
Detection Method Colorimetric
Quantity 1 Kit
Sample Type Body Fluid, Cell Culture Supernate, Serum, Tissue Lysate
Sample Volume 100 μL
Content And Storage Store at 4°C for 2 months, or at -20°C for 4 months.
For Use With (Application) Quantification
Product Type CTF1 (Human) ELISA Kit, Version 01
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.