Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ CTSB (Human) ELISA Kit

Catalog No. 89100731 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
Body Fluid, CCS, Plasma, Serum, Tissue Lysates
Body Fluid, CCS, Serum, Tissue Lysates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-100-731 Body Fluid, CCS, Plasma, Serum, Tissue Lysates
89-027-116 Body Fluid, CCS, Serum, Tissue Lysates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-100-731 Supplier Abnova™ Supplier No. KA3024
Add to Cart
Add to Cart

Sandwich Enzyme Immunoassay for the quantitative measurement of Human CTSB

Specifications

Calibration Range 156 to 10000pg/mL
Conjugate Biotin
Cross Reactivity Human
Description CTSB (Human) ELISA Kit is intended for the quantitative measurement of human CTSB.
Detection Limit 5pg/mL
Detection Method Colorimetric
Includes 96-well plate pre-coated with anti-Human Cathepsin B antibody, Lyophilized Human Cathepsin B standards: 10ng x 2, Sample / Standard diluent buffer: 30mL, Biotin conjugated anti-Human Cathepsin B antibody (Concentrated Dilution: 1:100): 130uL, Antibody diluent buffer: 12mL, Avidin-Biotin-Peroxidase Complex (ABC) (ConcentratedDilution: 1:100): 130uL, ABC diluent buffer: 12mL, TMB substrate: 10mL, Stop solution: 10mL, Wash buffer (25X): 30mL
Quantity 96 Assays
Sample Type Body Fluid, CCS, Plasma, Serum, Tissue Lysates
Sample Volume 100 μL
Content And Storage Store at 4°C for 4 months, or at -20°C for 8 months.
For Use With (Application) Quantification
Product Type CTSB (Human) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.