Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Cultured Cell Total RNA Purification Kit

Catalog No. 89028634 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-634 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-634 Supplier Abnova™ Supplier No. U0067
Add to Cart
Add to Cart

Designed specifically for purifying total RNA from cultured animal cells

  • Sample Type: Cultured cell
  • Sample Size: Cultured animal cells (up to 5 x 106)

Purification

Specifications

Content And Storage Store at room temperature.
Description Cultured Cell Total RNA Purification Kit was designed specifically for purifying total RNA from cultured animal cells.
For Use With (Application) Purification
Includes 60mL RB Buffer, 130mL W1 Buffer, 50mL Wash Buffer (add 200mL Ethanol), 30mL RNase-free Water, RNA binding plate, 0.35mL collection plate, adhesive film
Product Type Cultured Cell Total RNA Purification Kit
Quantity 1 Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.