Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Cxcl16 (Mouse) ELISA Kit

Catalog No. 89027120 Shop All Abnova Corporation Products
Change view
Click to view available options
Product Type:
CXCL16 (Mouse) ELISA Kit - Version 1
CXCL16 (Mouse) ELISA Kit - Version 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
89-027-120 CXCL16 (Mouse) ELISA Kit - Version 6
89-100-735 CXCL16 (Mouse) ELISA Kit - Version 1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-027-120 Supplier Abnova™ Supplier No. KA0556
Add to Cart
Add to Cart

Sandwich Enzyme Immunoassay for the quantitative measurement of Mouse Cxcl16

Specifications

Calibration Range 15.6 to 1000pg/mL
Conjugate Biotin
Cross Reactivity Mouse
Description Cxcl16 (Mouse) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of mouse Cxcl16.
Detection Limit 1pg/mL
Detection Method Colorimetric
Includes 96-well Antibody coated microwell, 10ng x 2 vials lyophilized recombinant Mouse CXcl16 standard, 30mL Sample Diluent Buffer, 130μL Biotinylated Anti-Mouse CXcl16 Antibody, 12mL Antibody Diluent Buffer, 130μL Avidin-Biotin-Peroxidase Complex(ABC), 12mL ABC diluent buffer, 10mL TMB color developing agent, 10mL TMB stop solution
Quantity 1 Kit
Sample Type Body Fluid, CCS, Plasma, Serum, Tissue Lysates
Sample Volume 100 μL
Content And Storage Store the kit at -20°C
For Use With (Application) Quantification
Product Type CXCL16 (Mouse) ELISA Kit - Version 6
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.