Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Cxcl16 (Mouse) ELISA Kit

Catalog No. 89100735 Shop All Abnova Corporation Products
Change view
Click to view available options
Product Type:
CXCL16 (Mouse) ELISA Kit - Version 1
CXCL16 (Mouse) ELISA Kit - Version 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
89-100-735 CXCL16 (Mouse) ELISA Kit - Version 1
89-027-120 CXCL16 (Mouse) ELISA Kit - Version 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-100-735 Supplier Abnova™ Supplier No. KA3028
Add to Cart
Add to Cart

Sandwich Enzyme Immunoassay for the quantitative measurement of Mouse Cxcl16

Specifications

Calibration Range 15.6 to 1000pg/mL
Conjugate Biotin
Cross Reactivity Mouse
Description Cxcl16 (Mouse) ELISA Kit is intended for the quantitative measurement of mouse Cxcl16.
Detection Limit 1pg/mL
Detection Method Colorimetric
Includes 96-well plate pre-coated with anti-Mouse Cxcl16 antibody, Lyophilized Cxcl16 standards (10ng x 2), Sample / Standard diluent buffer (30mL), Biotin conjugated anti-Mouse Cxcl16 antibody (Concentrated Dilution: 1:100) (130uL), Antibody diluent buffer (12mL), Avidin-Biotin-Peroxidase Complex (ABC) (ConcentratedDilution: 1:100) (130uL), ABC diluent buffer (12mL), TMB substrate (10mL), Stop solution (10mL), Wash buffer (25X) (30mL)
Quantity 96 Assays
Sample Type Body Fluid, CCS, Plasma, Serum, Tissue Lysates
Sample Volume 100 μL
Content And Storage Store at 4°C for 4 months, or at -20°C for 8 months.
For Use With (Application) Quantification
Product Type CXCL16 (Mouse) ELISA Kit - Version 1
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.