Learn More
Description
Includes
- Lyophilized recombinant mouse MIP-2 standard, 10ng x 2
- Plate precoated with anti-mouse MIP-2 antibody, 96-wells
- Sample diluent buffer, 30mL
- Biotinylated anti-mouse MIP-2 antibody, dilution 1:100, 130μL
- Antibody diluent buffer, 12mL
- Avidin-Biotin-Peroxidase complex (ABC), dilution 1:100, 130μL
- ABC diluent buffer, 12mL
- TMB color delevoping agent, 10mL
- TMB stop solution, 10mL
Specifications
Specifications
| Calibration Range | 15.6 to 1000pg/mL |
| Conjugate | HRP |
| Cross Reactivity | Mouse |
| Description | Cxcl2 (Mouse) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of mouse Cxcl2. |
| Detection Method | Colorimetric |
| Includes | 96-well Antibody coated microwell, 10ng x 2 vials lyophilized recombinant Mouse Cxcl2 standard, 30mL Sample Diluent Buffer, 130μL Biotinylated Anti-Mouse Cxcl2 Antibody, 12mL Antibody Diluent Buffer, 130μL Avidin-Biotin |
| Quantity | 1 Kit |
| Sample Type | Body Fluid, CCS, Plasma, Serum, Tissue Lysates |
| Sample Volume | 100 μL |
| Content And Storage | Store the kit at -20°C |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.