Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Cxcl2 (Mouse) ELISA Kit

Catalog No. 89100791 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
Body Fluid, CCS, Plasma, Serum, Tissue Lysates
Body Fluid, CCS, Serum, Tissue Lysates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-100-791 Body Fluid, CCS, Serum, Tissue Lysates
89-028-199 Body Fluid, CCS, Plasma, Serum, Tissue Lysates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-100-791 Supplier Abnova™ Supplier No. KA3088
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • Lyophilized recombinant mouse MIP-2 standard, 10ng x 2
  • Plate precoated with anti-mouse MIP-2 antibody, 96-wells
  • Sample diluent buffer, 30mL
  • Biotinylated anti-mouse MIP-2 antibody, dilution 1:100, 130μL
  • Antibody diluent buffer, 12mL
  • Avidin-Biotin-Peroxidase complex (ABC), dilution 1:100, 130μL
  • ABC diluent buffer, 12mL
  • TMB color delevoping agent, 10mL
  • TMB stop solution, 10mL

Sandwich Enzyme Immunoassay for the quantitative measurement of Mouse Cxcl2

Specifications

Calibration Range 15.6 to 1000pg/mL
Description Cxcl2 (Mouse) ELISA Kit is intended for the quantitative measurement of mouse Cxcl2.
Detection Method Colorimetric
Quantity 1 Kit
Sample Type Body Fluid, CCS, Serum, Tissue Lysates
Sample Volume 100 μL
Content And Storage Store at 4°C for 4 months, or at -20°C for 8 months.
For Use With (Application) Quantification
Product Type Cxcl2 (Mouse) ELISA Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.