Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Cyclic AMP Complete ELISA Kit

Catalog No. 89100180 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-180 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-180 Supplier Abnova™ Supplier No. KA0320
Add to Cart
Add to Cart

Includes

  • Assay Buffer 2, 27mL
  • 0.1M HCl, 27mL
  • cyclic AMP Standard, 0.5mL
  • Triethylamine, 2mL
  • Acetic Anhydride, 1mL
  • Goat anti-Rabbit IgG Microtiter Plate, One plate of 96 wells
  • Neutralizing Reagent, 5mL
  • cAMP Antibody, 5mL
  • cAMP Conjugate, 5mL
  • Wash Buffer Concentrate, 30mL
  • pNpp Substrate, 20mL
  • Stop Solution, 5mL
  • Plate Sealer, 1 slice

Use to measure quantitatively cyclic AMP in cells and tissue treated with 0.1M HCl, in addition to culture supernates, saliva, and serum. A kit.

The cyclic AMP Complete ELISA Kit is a competitive immunoassay for quantitative determination of cyclic AMP in cells and tissue treated with 0.1M HCl, in addition to culture supernates, saliva, and serum

Specifications

Calibration Range 0.078 to 20pmol/mL
Conjugate AP
Description cyclic AMP Complete ELISA Kit is a kit for the quantitative determination of cyclic AMP in cells and tissue treated with 0.1M HCl, in addition to culture supernates, saliva, and serum.
Detection Limit Assay Buffer: 0.30pmol/mL (non-acetylated); 0.039pmol/mL (acetylated); HCl: 0.39pmol/mL (non-acetylated); 0.037pmol/mL (acetylated)
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status RUO
Sample Type Cell Culture Supernatant, Cell Lysate, Saliva, Serum, Tissue
Sample Volume 100 μL
Sensitivity 0.30 pmol/mL in the non-acetylated assay format and 0.039 pmol/mL in the acetylated assay format
Packaging 96-well
Content And Storage The standard and conjugate must be stored at -20°C; others store at 4°C.
For Use With (Application) Quantification
Product Type cyclic AMP Complete ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.