Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Cyclic GMP Complete ELISA Kit

Catalog No. 89100179 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-179 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-179 Supplier Abnova™ Supplier No. KA0321
Only null left
Add to Cart
Add to Cart

Includes

  • Assay Buffer 2, 27mL
  • 0.1M HCl, 27mL
  • cyclic GMP Standard, 0.5mL
  • Triethylamine, 2mL
  • Acetic Anhydride, 1mL
  • Goat anti-Rabbit IgG Microtiter Plate, One plate of 96 wells
  • Neutralizing Reagent, 5mL
  • cGMP Antibody, 5mL
  • cGMP Conjugate, 5mL
  • Wash Buffer Concentrate, 27mL
  • pNpp Substrate, 20mL
  • Stop Solution, 5mL
  • Plate Sealer, 1 each

Use to measure quantitatively cyclic GMP in cells and tissue treated with 0.1N HCl, in addition to culture supernates, saliva, and serum. A kit.

Specifications

Calibration Range 0.08 to 50pmol/mL
Conjugate AP
Description cyclic GMP Complete ELISA Kit is a kit for the quantitative determination of cyclic GMP in cells and tissue treated with 0.1N HCl, in addition to culture supernates, saliva, and serum.
Detection Limit Assay Buffer: 0.42pmol/mL (non-acetylated); 0.043pmol/mL (acetylated) HCl: 0.604pmol/mL (non-acetylated); 0.059pmol/mL (acetylated)
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Cell Culture Supernatant, Cell Lysate, Plasma, Saliva, Serum, Tissue, Urine
Sample Volume 100 μL
Sensitivity Assay Buffer 3: 0.42 pmol/mL (non-acetylated); 0.043 pmol/mL (acetylated)0.1M HCl: 0.604 pmol/mL (non-acetylated); 0.059 pmol/mL (acetylated)
Packaging 96-well
Content And Storage The standard and conjugate must be stored at -20°C; others store at 4°C.
For Use With (Application) Quantification
Product Type cyclic GMP Complete ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.