Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Cysteinyl Leukotriene ELISA Kit

Catalog No. 89100189 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-189 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-189 Supplier Abnova™ Supplier No. KA0311
Add to Cart
Add to Cart

Includes

  • Goat anti-Mouse IgG Microtiter Plate, 96 Wells
  • Cysteinyl Leukotriene ELISA Conjugate, 6mL
  • Cysteinyl Leukotriene ELISA Antibody, 6mL
  • Assay Buffer, 30mL
  • Wash Buffer Concentrate, 30mL
  • Cysteinyl Leukotriene Standard, 0.5mL
  • pNpp Substrate, 20mL
  • Stop Solution, 6mL
  • Plate Sealer, 1 slice

Use to measure quantitatively Cysteinyl Leukotrienes in tissue culture media and urine. A competitive immunoassay.

The kit measures three of the major metabolites of LTA4; LTC4, LTD4 and LTE4 and uses a monoclonal antibody to the Cysteinyl Leukotrienes (LTC4, LTD4 and LTE4) to bind, in a competitive manner, the Cysteinyl Leukotrienes in the sample or an alkaline phosphatase molecule which has LTC4 covalently attached to it

Specifications

Calibration Range 78.1 to 2500pg/mL
Conjugate AP
Description Cysteinyl Leukotriene ELISA Kit is a competitive immunoassay for the quantitative determination of Cysteinyl Leukotrienes in tissue culture media and urine.
Detection Limit 26.6pg/mL
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Tissue culture media, urine
Sample Volume 100 μL
Sensitivity 26.6 pg/mL
Packaging 96-well
Content And Storage The standard and conjugate must be stored at -20°C; others store at 4°C.
For Use With (Application) Quantification
Product Type Cysteinyl Leukotriene ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.