Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ CytoQuest™ CR: Bottle and Tubing Pack

Catalog No. 89139312 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-139-312 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-139-312 Supplier Abnova™ Supplier No. KA4442
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Each pack contains one each of: 70% EtOH Bottle, DI Water Bottle, PBS Bottle, Wash Medium Bottle, Fixation Bottle, 70% EtOH Line (Green), DI Water Line (Yellow), PBS Line (Blue), Wash Medium Line (Red), Fixation Line, Sample Line, Waste Line, Air Filter

Bottle and Tubing Pack contains fluidic bottles and tubings for periodical replacement on CytoQuest™ CR

Applications:
Immunofluorescence

Recommended Storage

Store at room temperature.

Specifications

Content And Storage Store at room temperature.
Includes Each pack contains one each of: 70% EtOH Bottle, DI Water Bottle, PBS Bottle, Wash Medium Bottle, Fixation Bottle, 70% EtOH Line (Green), DI Water Line (Yellow), PBS Line (Blue), Wash Medium Line (Red), Fixation Line, Sample Line, Waste Line, Air Filter
Quantity 1 Kit
For Use With (Equipment) CytoQuest CR

For research use only (RUO).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.