Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ CytoQuest™ SuperBlocker

Catalog No. 89139601 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-139-601 200μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-139-601 Supplier Abnova™ Supplier No. U0317
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

10X CytoQuest SuperBlocker

Inhibits non-specific binding of RBC and WBC to the CytoChipNano and CytoChipNano CR

  • Reagent is sufficient for 20 assays using recommended protocol
  • Quantity: 200μL

Applciations:
Immunofluorescence

Recommended Storage

Store at 4°C. Aliquot to avoid repeated freezing and thawing.

Specifications

Content And Storage Store at 4°C. Aliquot to avoid repeated freezing and thawing.
Includes 10X CytoQuest SuperBlocker
Quantity 200μL
For Use With (Equipment) CytoQuest CR

For research use only (RUO).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.