Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ CytoQuest™ Trophoblast Kit

Catalog No. 89139343 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-139-343 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-139-343 Supplier Abnova™ Supplier No. KA4515
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Includes

  • Anti-HLA-G capturing antibody (Biotin): Biotin conjugated Anti-HLA-G antibody for trophoblast capturing.
  • Anti-TBA detecting antibody (TRITC): TRITC conjugated Anti-TBA antibody for trophoblast detection.
  • FcR Blocker
  • 1X TADB

CytoQuest Trophoblast Kit contains antibodies for immobilization and immunostaining of circulating circulating trophoblasts

  • Reagents are sufficient for 20 assays using recommended protocol
Applications:
Immunofluorescence

Recommended Storage

Store at 4°C. Aliquot to avoid repeated freezing and thawing.

Specifications

Content And Storage Store at 4°C. Aliquot to avoid repeated freezing and thawing.
Includes
  • Anti-HLA-G capturing antibody (Biotin): Biotin conjugated Anti-HLA-G antibody for trophoblast capturing.
  • Anti-TBA detecting antibody (TRITC): TRITC conjugated Anti-TBA antibody for trophoblast detection.
  • FcR Blocker
  • 1X TADB
Quantity 1 Kit
For Use With (Equipment) CytoQuest CR

For research use only (RUO).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.