Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Dengue virus IgG ELISA Kit

Catalog No. 89028058 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
96 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-058 96 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-058 Supplier Abnova™ Supplier No. KA0961
Add to Cart
Add to Cart

Includes

  • Microwells coated with Dengue antigen, 12 x 8 x 1
  • Sample Diluent: 1 bottle (ready to use), 22mL
  • Calibrator: yellow Cap. 1 Vial (ready to use), 1mL
  • Positive Control: Red Cap. 1 vial (ready to use) 1mL
  • Negative Control: Blue Cap. 1 vial (ready to use) 1mL
  • Enzyme conjugate: 1 bottle (ready to use) 12mL
  • TMB Substrate: 1 bottle (ready to use) 12mL
  • Stop Solution: 1 bottle (ready to use) 12mL
  • Wash concentrate 20X: 1 bottle 25mL

For the detection of IgG antibody to Dengue Virus in Human Serum or Plasma

Specifications

Conjugate HRP
Description Dengue virus IgG ELISA Kit is used for the detection of IgG antibody to Dengue virus in human serum or plasma.
Detection Method Colorimetric
Includes 12 x 8 x 1 Microwell coated with Dengue Antigen, 22mL sample diluent, 1.5mL Calibrator, 1.5mL positive control, 1.5mL negative control, 12mL Enzyme Conjugate, 12mL TMB Substrate, 12mL stop solution, 25mL (20X) Wash concentrate 1 bottle
Quantity 96 Assays
Sample Type Plasma, Serum
Sample Volume 10 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Qualitative
Product Type Dengue virus IgG ELISA Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.