Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ DHEA ELISA Kit

Catalog No. 89100185 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-185 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-185 Supplier Abnova™ Supplier No. KA0315
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • Goat anti-Rabbit IgG Microtiter Plate, 96 Wells
  • DHEA ELISA Conjugate, 5mL
  • DHEA ELISA Antibody, 5mL
  • Assay Buffer, 27mL
  • Wash Buffer Concentrate, 27mL
  • DHEA Standard, 0.5mL
  • p-Npp Substrate, 20mL
  • Stop Solution, 5mL
  • Plate Sealer, 2 slides

Use to measure quantitatively DHEA in biological fluids. A competitive immunoassay.

The DHEA kit is a competitive immunoassay for the quantitative determination of DHEA in biological fluids

Specifications

Calibration Range 12.21 to 50,000pg/mL
Conjugate AP
Description DHEA ELISA Kit is a competitive immunoassay for the quantitative determination of DHEA in biological fluids.
Detection Limit 2.9pg/mL
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status RUO
Sample Type Cell Culture Supernatant, Plasma, Saliva, Serum, Urine
Sample Volume 100 μL
Sensitivity 2.90 pg/mL
Packaging 96-well
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type DHEA ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.