Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ DHEA-S ELISA Kit

Catalog No. 89028718 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-718 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-718 Supplier Abnova™ Supplier No. KA1902
Add to Cart
Add to Cart

Enzyme Immunoassay for the quantitative measurement of free DHEA-S in Saliva

Specifications

Conjugate HRP
Description DHEA-S ELISA Kit is an enzyme immunoassay for the quantitative measurement of free DHEA-S in saliva.
Detection Limit 0.045ng/mL
Detection Method Colorimetric
Includes 1mL Standard A, 1mL standard B, 1mL standard C, 1mL standard D, 1mL standard E, 30mL INC Buffer, 0.4mL Conjugated conc, 96-well microplate, 12mL Substrate, 12mL stop solution, 20mL (50X) Wash-Conc
Quantity 1 Kit
Sample Type Saliva
Sample Volume 50 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type DHEA-S ELISA Kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.