Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ DHEA-S (Human) ELISA Kit

Catalog No. 89024461 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
96 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-024-461 96 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-024-461 Supplier Abnova™ Supplier No. KA0920
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For the quantitative measurement of DHEA-S in Human Serum or Plasma

Specifications

Calibration Range 0.1 to 10ng/mL
Conjugate HRP
Cross Reactivity Human
Description DHEA-S (Human) ELISA Kit is used for the quantitative measurement of DHEA-S in human serum or plasma.
Detection Limit 0.02μg/mL
Detection Method Colorimetric
Includes 12 x 8 x1 Microwell strip, 25mL Enzyme Conjugate, 1mL x 7 vials positive reference standard, 14mL TMB Chromogenic Substrate, 14mL stop solution, 30mL (40X) Wash solution
Quantity 96 Assays
Sample Type Plasma, Serum
Sample Volume 25 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type DHEA-S (Human) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.