Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Diacetylspermine ELISA Kit

Catalog No. 89028135 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-135 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-135 Supplier Abnova™ Supplier No. KA1002
Add to Cart
Add to Cart

Sandwich Enzyme Immunoassay for the quantitative measurement of Diacetylspermine

Specifications

Calibration Range 6.25 to 200nm
Conjugate HRP
Cross Reactivity Human
Description Diacetylspermine ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of diacetylspermine.
Detection Method Colorimetric
Includes 96-well-Antigen coated microtiter plate, 2 250μL Diacetylspermine standard, 20mL Antibody Diluent, 60μLl anti Diacetylspermine Antibody(100X), 80μL HRP-anti Rabbit IgG Antibody(80x), 2 o-phenylendiamin
Quantity 1 Kit
Sample Type Urine
Sample Volume 50 μL
Content And Storage Store the kit at -20°C
For Use With (Application) Quantification
Product Type Diacetylspermine ELISA kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.