Learn More
Description
- The DNA Demethylase Activity/Inhibition Assay Kit is a complete set of optimized reagents for measuring DNA demethylase activity or inhibition.
- Application: DNA demethylase activity/inhibition assays
- Format: Colorimetric detection
- Storage: Suitable for use in standard laboratory settings
Specifications
Specifications
| Description | DNA Demethylase Activity/Inhibition Assay Kit is use for measuring DNA demethylase activity/inhibition. |
| Detection Method | Colorimetric |
| Includes | 22mL Wash Buffer (10X) (DD1), 3mL Demethylase Assay Buffer (DD2), 0.2mL Demethylase Substrate (DD3), 40μL Demethylation standard (DD4), 8μL capture Antibody (DD5), 20μL detecting Antibody (DD6), 12mL d |
| Quantity | 1 Kit |
| Content And Storage | Store DD3, DD4 and DD6 at -20°C. Store DD5, DD7, and 8 well strips at 4°C away from light. Store other components at room temperature. |
| Form | Lyophilized |
| For Use With (Application) | Functional Study |
| Product Type | DNA Demethylase activity/inhibition assay kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.