Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ DNA Methyltransferase Activity/Inhibition Assay Kit (Fluorometric)

Catalog No. 89027038 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-027-038 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-027-038 Supplier Abnova™ Supplier No. KA0681
Add to Cart
Add to Cart

For measuring Dnmt activity/inhibition

Specifications

Description DNA Methyltransferase Activity/Inhibition Assay Kit (Fluorometric) is use for measuring Dnmt activity/inhibition.
Detection Method Fluorometric
Includes 22mL Wash Buffer (10X) (MT1), 3mL DNMT Assay Buffer (MT2), 70μL adomet (MT3), 10μL DNMT positive control (MD4), 8μL capture Antibody (MT5), 20μL detecting Antibody (MT6), 24
Quantity 1 Kit
Content And Storage Store MD3, MD4, MD6, and MD7 at -20°C away from light. Store all other components at 4°C away from light.
For Use With (Application) Functional Study
Product Type DNA Methyltransferase activity/inhibition assay kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.