Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ DPP4 (Human) ELISA Kit

Catalog No. 89101014 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-101-014 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-101-014 Supplier Abnova™ Supplier No. KA3335
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • 96-well plate precoated with anti-human CD26 antibody, 96 (8 x 12) wells
  • Lyophilized recombinant human CD26 standard, 10ng x 2
  • Biotinylated anti-human CD26 antibody (dilution 1:100), 130μL
  • Avidin-Biotin-Peroxidase Complex (ABC) (dilution 1:100), 130μL
  • Sample diluent buffer, 30mL
  • Antibody diluent buffer, 12mL
  • ABC diluent buffer, 12mL
  • TMB color developing agent, 10mL
  • TMB stop solution, 10mL

Use to measure quantitatively human DPP4. Sandwich enzyme immunoassay.

DPP4 (Human) ELISA Kit is based on standard sandwich enzyme-linked immune-sorbent assay technology

Specifications

Calibration Range 0.312 to 20ng/mL
Conjugate Biotin
Cross Reactivity Human
Description DPP4 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human DPP4.
Detection Limit 10pg/mL
Detection Method Colorimetric
Includes 96-well plate precoated with anti- human CD26 antibody (96 (8 x 12) wells), Lyophilized recombinant human CD26 standard (10ng x 2), Biotinylated anti- human CD26 antibody(dilution 1:100); 130μL, Avidin-Biotin-Peroxidase Complex (ABC) (dilution 1:100): 130μL, Sample diluent buffer (30mL), Antibody diluent buffer (12mL), ABC diluent buffer (12mL), TMB color developing agent (10mL), TMB stop solution (10mL)
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Cell Culture Supernate, Plasma (EDTA, Heparin), Saliva, Serum, Urine
Sample Volume 100 μL
Sensitivity <10 pg/mL
Packaging 96-well
Content And Storage Store the kit at -20°C
For Use With (Application) Quantification
Product Type DPP4 (Human) ELISA Kit
Specificity Natural and recombinant human CD26
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.