Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ dsDNA IgG ELISA Kit

Catalog No. 89027238 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
96 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-027-238 96 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-027-238 Supplier Abnova™ Supplier No. KA1100
Add to Cart
Add to Cart

Indirect solid phase ELISA for the quantitative measurement of IgG class autoantibodies against double-stranded DNA in Human Serum or Plasma

Specifications

Calibration Range 12.5 to 200IU/mL
Conjugate HRP
Description ds-DNA Ab IgG ELISA Kit is an indirect solid phase ELISA for the quantitative measurement of IgG class autoantibodies against double-stranded DNA in human serum or plasma.
Detection Method Colorimetric
Includes 6 1.5mL combined calibrator with IgG class Anti-dsDNA Antibody vials, 2 1.5mL Anti-dsDNA control vials, one 20mL Sample Buffer vial, one 15mL Enzyme Conjugate solution vial, one 15mL TMB Substrate solution vial, one 15mL stop solution vial, one 96-well plate, one 20mL wash solution vial
Quantity 96 Assays
Sample Type Plasma, Serum
Sample Volume 10 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type dsDNA IgG ELISA Kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.