Learn More
Specifications
Specifications
| Calibration Range | 12.5 to 200U/mL |
| Conjugate | HRP |
| Description | dsDNA Screen ELISA Kit is an indirect solid phase ELISA for the quantitative measurement of IgG, IgM and IgA class autoantibodies against double-stranded DNA in human serum or plasma. |
| Detection Method | Colorimetric |
| Includes | 6 1.5mL combined calibrator with class Anti-dsDNA Antibody vials, 2 1.5mL Anti-dsDNA control vials, one 20mL Sample Buffer vial, one 15mL Enzyme Conjugate solution vial, one 15mL TMB Substrate solution vial, 15mL stop solution vial, one 96-well plate, one 20mL wash solution vial |
| Quantity | 96 Assays |
| Sample Type | Plasma, Serum |
| Sample Volume | 10 μL |
| Content And Storage | Store the kit at 4°C |
| For Use With (Application) | Quantification |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.