Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ EGFR (pY1068)/Pan EGFR (Human) ELISA Kit

Catalog No. 89100438 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-438 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-438 Supplier Abnova™ Supplier No. KA2158

missing translation for 'validMainframeMsg'

Includes

  • EGFR Microplate
  • Wash Buffer Concentrate (20x)
  • Anti-phospho-EGFR (Tyr 1068): 11μL of rabbit anti-human EGFR (Tyr 1068)
  • HRP-conjugated Anti-rabbit IgG, 25μL of HRPconjugated Anti-rabbit IgG
  • Biotinylated-Anti-EGFR: 1 vial goat anti-human EGFR
  • HRP-Streptavidin concentrate: 8μL of 30,000 fold concentrated HRP-Streptavidin concentrate
  • Assay Diluent: 15mL of 5x concentrated buffer. For diluting cell lysate, Biotinylated antibody and HRPStreptavidin diluent
  • TMB One-Step Substrate Reagent
  • Stop Solution: 2M sulfuric acid
  • Cell Lysate Buffer
  • Positive Control A431S002-1: cell lysate powder

Use to detect qualitatively human phospho EGFR (Y1068) and Pan EGFR. Enzyme-linked immunosorbent assay.

EGFR (pY1068)/Pan EGFR (Human) ELISA (Enzyme-Linked Immunosorbent Assay) kit is a very sensitive assay kit that can monitor the activation or function of important biological pathways in cell lysates

Specifications

Conjugate HRP
Cross Reactivity Human
Description EGFR (pY1068)/Pan EGFR (Human) ELISA Kit is an enzyme-linked immunosorbent assay for the qualitative determination of human phospho EGFR (Y1068) and Pan EGFR.
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Cell Lysate, Tissue Lysate
Sample Volume 100 μL
Packaging 96-well
Content And Storage Store the kit at -20°C
For Use With (Application) Qualitative
Product Type EGFR (pY1068)/Pan EGFR (Human) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.