Learn More
Specifications
Specifications
| Conjugate | HRP |
| Description | ENA-6-Profile ELISA Kit is an indirect solid phase ELISA for semi-quantitative determination of IgG class autoantibodies directed against the extractable nuclear antigens SS-A (Ro), SS-B (La), Sm RNP/Sm, Scl-70 and Jo-1 in human serum or plasma. |
| Detection Method | Colorimetric |
| Includes | 1 x 2mL Positive control vial, 1 x 2mL negative control vial, 6 x 1mL Cut-off calibrator, one 20mL Sample Buffer, one 15mL Enzyme Conjugate solution, one 15mL TMB Substrate solution, one 15mL stop solution, one 96-well plate, one 20mL wash solution |
| Quantity | 96 Tests |
| Sample Type | Plasma, Serum |
| Sample Volume | 10 μL |
| Content And Storage | Store the kit at 4°C |
| For Use With (Application) | Quantification |
| Product Type | ENA-6 Profile ELISA Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.