Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ ENA Combi ELISA Kit

Catalog No. 89027241 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-027-241 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-027-241 Supplier Abnova™ Supplier No. KA1103
Add to Cart
Add to Cart

Indirect solid phase ELISA for the semi-quantitative measurement of IgG class autoantibodies against extractable nuclear antigens (ENA) in Human Serum or Plasma

Specifications

Calibration Range 12.5 to 100U/mL
Conjugate HRP
Description ENA Combi ELISA Kit is an indirect solid phase ELISA for the semi-quantitative measurement of IgG class autoantibodies against extractable nuclear antigens (ENA) in human serum or plasma.
Detection Method Colorimetric
Includes 1.5mL Anti-ENA control vial, 20mL Sample Buffer, 15mL Enzyme Conjugate solution, 15mL TMB Substrate solution, 15mL stop solution, 20mL wash solution, one 96-well plate
Quantity 96 Tests
Sample Type Plasma, Serum
Sample Volume 10 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type ENA Combi ELISA Kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.