Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ ENA Screen ELISA Kit

Catalog No. 89027244 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-027-244 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-027-244 Supplier Abnova™ Supplier No. KA1106
Add to Cart
Add to Cart

Indirect solid phase ELISA for the qualitative screening of IgG class autoantibodies against extractable nuclear antigens (ENA) in Human Serum or Plasma

Specifications

Conjugate HRP
Description ENA Screen ELISA is an indirect solid phase ELISA for the screening of IgG class autoantibodies against extractable nuclear antigens (ENA) in human serum or plasma.
Detection Method Colorimetric
Includes 3 1.5mL Anti-ENA Controls in a Serum/Buffer Matrix vials, one 20mL Sample Buffer, one 15mL Enzyme Conjugate solution, one 15mL TMB Substrate solution, one 15mL stop solution, one 96-well plate, one 20mL wash solution vial
Quantity 96 Tests
Sample Type Plasma, Serum
Sample Volume 10 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Qualitative
Product Type ENA Screen ELISA Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.