Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Epinephrine ELISA Kit

Catalog No. 89028615 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-615 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-615 Supplier Abnova™ Supplier No. KA1882
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Enzyme Immunoassay for the quantitative determination of Epinephrine in Plasma and Urine

Specifications

Calibration Range 4.6 to 81.4ng/mL Urine, 92 to 545pg/mL Plasma
Description Epinephrine ELISA Kit is an enzyme immunoassay for the quantitative determination of Epinephrine in plasma and urine.
Detection Method Colorimetric
Includes Adhesive foil 1 x 4, 20mL Wash Buffer concentrate, 12mL Enzyme Conjugate, 12mL Substrate, 12mL stop solution, 96-Adrenaline-Metanephrine microtiter strip well, 6mL Adrenaline antiserum, 4mL Adjustment Buffer, 4mL standard A, 4mL standard B, 4mL standard C, 4mL standard D, 4mL standard E, 4mL standard F, 20mL Acylation Buffer, 3mL Acylation reagent, 6mL Assay Buffer, 2mL coenzyme, 1mL Enzyme, 6mL Extraction Buffer, 2 48 extraction plate, 20mL Hydrochloric acid, 4mL control 1, 4mL control 2
Quantity 1 Kit
Sample Type Plasma, Urine
Sample Volume 50 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type Epinephrine ELISA kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.