Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Epinephrine/Norepinephrine/Dopamine ELISA Kit

Catalog No. 89028613 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-613 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-613 Supplier Abnova™ Supplier No. KA1880
Add to Cart
Add to Cart

Includes

  • Adhesive foil ready for use, 4 x 3
  • Wash buffer Concentrate, Concentrate. Dilute content with dist. Water to a final volume of 1000mL, 20mL x 3
  • Enzyme Conjugate ready for use, anti-rabbit IgG conjugated with peroxidase, 12mL x 3
  • Substrate ready for use, containing a solution of tetramethylbenzidine (TMB), 12mL x 3
  • Stop solution ready for use, containing 0.25M H2SO4, 12mL x 3
  • Adrenaline-Metanephrine microtiter strips, 12 strips, 8 wells each, break apart, precoated, blue coloured, 96 ( 8 x 12) wells
  • Noradrenaline-Normetanephrine microtiter strips 12 strips, 8 wells each, break apart, precoated, yellow coloured, 96 (8 x 12) wells
  • Dopamine microtiter Strips 12 strips, 8 wells each, break apart, precoated, green coloured, 96 (8 x 12) wells
  • Adrenaline antiserum from rabbit, ready for use, blue coloured, blue screw cap, 6mL

Enzyme Immunoassay for the quantitative determination of Epinephrine, Norepinephrine and Dopamine in Plasma and Urine

Specifications

Calibration Range Epinephrine: 1.3 to 81.4ng/mL, Norepinephrine: 4.9 to 339ng/mL, Dopamine: 14 to 917ng/mL
Description Epinephrine/Norepinephrine/Dopamine ELISA Kit is an enzyme immunoassay for the quantitative determination of Epinephrine, Norepinephrine and Dopamine in plasma and urine. This product contains three assay plates, for Epinephrine, Norepinephrine and Dopamine separately.
Detection Method Colorimetric
Includes Adhesive foil 4 x 3, 3 x 20mL Wash Buffer concentrate, three 12mL Enzyme Conjugate, three 12mL Substrate, three 12mL stop solution, 96-Adrenaline-Metanephrine microtiter strip, 96-Noradrenaline-Normetanephrine Microtiter strip, 96 Dopamine microtiter strip, 6mL Adrenaline antiserum, 6mL Noradrenaline antiserum, 6mL Dopamine, antiserum, 2 4mL adjustment Buffer, 4mL standard A, 4mL standard B, 4mL standard C, 4mL standard D, 4mL standard E, 4mL Standard F, 20mL Acylation Buffer, 3ml Acylationreagent, 6mL Assay Buffer, 4mL coenzyme, six 1mL Enzyme, 6mL Extraction Buffer, 2 48-well extraction plate, 20mL Hydrochloric acid, 4mL control 1, 4mL control 2, 4mL standard A/B
Quantity 1 Kit
Sample Type Plasma, Urine
Sample Volume Urine: 10 μL, Plasma: 300 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type Epinephrine/Norepinephrine/Dopamine ELISA kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.