Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Epstein Barr Virus EBNA-1 IgM ELISA Kit

Catalog No. 89028289 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-289 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-289 Supplier Abnova™ Supplier No. KA1449
Add to Cart
Add to Cart

Semiquantitative measurement of IgM autoantibodies against Epstein Barr Virus EBNA-1

Specifications

Description Epstein Barr Virus EBNA-1 IgM ELISA Kit is a solid phase enzyme immunoassay for the semiquantitative measurement of IgM autoantibodies against Epstein Barr Virus EBNA-1.
Detection Method Colorimetric
Includes 96 ELISA break away strip coated with specific Antigen, 1.3mL standard A to F vials, 15mL Antibodies-Px Conjugate, 125mL Wash Buffer concentrate, 125mL Dilution Buffer, 15mL chromogenic Substrate, 15mL stop solution
Quantity 1 Kit
Sample Type Plasma, Serum
Sample Volume 100 μL
Content And Storage Store at 4°C on dry atmosphere away from light.
For Use With (Application) Quantification
Product Type Epstein Barr Virus EBNA-1 IgM ELISA kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.