Learn More
Specifications
Specifications
| Conjugate | HRP |
| Description | Epstein Barr Virus VCA IgG ELISA Kit is a solid phase enzyme immunoassay for the quantitative measurement of IgG class autoantibodies against Epstein Barr Virus VCA. |
| Detection Method | Colorimetric |
| Includes | 96 ELISA break away strip coated with specific Antigen, 1.3mL standard A to F, 15mL Antibodies-Px Conjugate, 125mL Wash Buffer concentrate, 125mL Dilution Buffer, 15mL chromogenic Substrate, 30mL stop solution |
| Quantity | 1 Kit |
| Regulatory Status | For research use only (RUO) |
| Sample Type | CSF, Serum |
| Sample Volume | 100 μL |
| Content And Storage | Store at 4°C on dry atmosphere away from light. |
| For Use With (Application) | Quantification |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.