Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Epstein Barr Virus VCA IgG ELISA Kit

Catalog No. 89100516 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
CSF, Serum
Plasma, Serum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-100-516 Plasma, Serum
89-028-285 CSF, Serum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-100-516 Supplier Abnova™ Supplier No. KA2254
Add to Cart
Add to Cart

Solid phase Enzyme Immunoassay for the quantitative measurement of IgG class autoantibodies against Epstein Barr Virus VCA

Specifications

Conjugate HRP
Description Epstein Barr Virus VCA IgG ELISA Kit is intended for the detection of IgG antibody to EBV-VCA in human serum or plasma.
Detection Method Colorimetric
Includes Component (Size), Microwell strips (12 x 8 x 1 ), Sample Diluent (22mL), Calibrator (1.5mL), Positive Control (1.5mL), Negative Control (1.5mL), Enzyme Conjugate (12mL), TMB Chromogenic Substrate (12mL), Stop Solution (12mL), Wash Concentrate 20x (25mL)
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Plasma, Serum
Sample Volume 10 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Qualitative
Product Type Epstein Barr Virus VCA IgG ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.