Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ ERBB2 (Human) ELISA Kit

Catalog No. 89027208 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
Body Fluid, CCS, Plasma, Serum, Tissue Lysates
CCS, Plasma (EDTA, Heparin), Serum, Tissue Lysates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-027-208 CCS, Plasma (EDTA, Heparin), Serum, Tissue Lysates
89-100-741 Body Fluid, CCS, Plasma, Serum, Tissue Lysates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-027-208 Supplier Abnova™ Supplier No. KA1070
Add to Cart
Add to Cart

Sandwich Enzyme Immunoassay for the quantitative measurement of Human ERBB2

Specifications

Calibration Range 62.5 to 4000pg/mL
Conjugate Biotin
Cross Reactivity Human
Description Human ERBB2 ELISA kit is a sandwich enzyme immunoassay for the quantitative measurement of human ERBB2.
Detection Limit 10pg/mL
Detection Method Colorimetric
Includes 96 Plate precoated with Anti-Human ErbB-2 Antibody well, 10ng lyophilized recombinant Human ErbB-2 standard, 130μL Biotinylated Anti-Human ErbB-2 Antibody, 130μL Avidin-Biotin-Peroxidase Complex (ABC), TMB substrate (10mL), Stop solution (10mL), Wash buffer (25X) (30mL)
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type CCS, Plasma (EDTA, Heparin), Serum, Tissue Lysates
Sample Volume 100 μL
Content And Storage Store the kit at -20°C
For Use With (Application) Quantification
Product Type ERBB2 (Human) ELISA kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.